siRNA Details
Gene Symbol: Tsen15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025677.3 | NP_079953.2 | 4 | 557 | ||
GenBank mRNA | AK004261.1 | - | 4 | 493 | ||
AK011608.1 | - | 4 | 514 | |||
AK017650.1 | - | 4 | 558 | |||
AK169429.1 | - | 3 | 3312 | |||
BC024342.1 | - | 4 | 487 | 5002659 | ||
BC145701.1 | - | 4 | 524 | 40131004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
