siRNA Details
Gene Symbol: Sod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011434.1 | NP_035564.1 | 2 | 224 | ||
GenBank mRNA | AK020624.1 | - | 2 | 130 | ||
AK077284.1 | - | 2 | 164 | |||
AK080908.1 | - | 1 | 2069 | |||
AK080929.1 | - | 1 | 2068 | |||
BC002066.1 | - | 2 | 178 | 3496016 | ||
BC048874.1 | - | 2 | 224 | 5401293 | ||
BC057074.1 | - | 1 | 2087 | |||
BC086886.1 | - | 2 | 216 | 6760550 | ||
M35725.1 | - | 2 | 112 | |||
X06683.1 | - | 2 | 112 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
