siRNA Details
Gene Symbol: Gm5886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177652.2 | NP_001171123.1 | 3 | 436 | ||
GenBank mRNA | AB256662.1 | - | 3 | 423 | ||
BC019499.1 | - | 2 | 1215 | |||
BC038040.1 | - | 2 | 1234 | |||
BC055452.1 | - | 3 | 438 | |||
BC055924.1 | - | 3 | 424 | |||
BC069936.1 | - | 3 | 424 | |||
BC087541.1 | - | 3 | 431 | |||
BC108376.1 | - | 3 | 430 | |||
BG865695.1 | - | 3 | 435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
