siRNA Details
Gene Symbol: Crxos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033638.2 | NP_001028810.1 | 5 | 662 | ||
NM_001145190.2 | NP_001138662.1 | 3 | 299 | |||
GenBank mRNA | AB472692.1 | - | 5 | 615 | ||
AB472694.1 | - | 3 | 283 | |||
AK145701.1 | - | 5 | 662 | |||
AY590890.1 | - | 3 | 356 | |||
AY590891.1 | - | 4 | 530 | |||
CJ040988.1 | - | 3 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
