siRNA Details
Gene Symbol: Mast3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199308.2 | NP_955012.2 | 7 | 855 | ||
GenBank mRNA | AK140821.1 | - | 7 | 862 | ||
CB524773.1 | - | 3 | 289 |

More Information
Other Species Targeted

Gene Ontology Categories:
