siRNA Details
Gene Symbol: Ube2i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177609.1 | NP_001171080.1 | 7 | 1189 | ||
NM_001177610.1 | NP_001171081.1 | 6 | 478 | |||
NM_011665.4 | NP_035795.1 | 6 | 516 | |||
GenBank mRNA | AK003141.1 | - | 6 | 423 | ||
AK005058.1 | - | 6 | 498 | |||
AK011239.1 | - | 6 | 424 | |||
AK012282.1 | - | 6 | 442 | |||
AK088508.1 | - | 6 | 501 | |||
AK150575.1 | - | 6 | 459 | |||
AK160014.1 | - | 6 | 445 | |||
AK160988.1 | - | 6 | 442 | |||
AK165606.1 | - | 6 | 428 | |||
AK165615.1 | - | 6 | 446 | |||
AK166016.1 | - | 6 | 426 | |||
AK166657.1 | - | 7 | 1190 | |||
AK166930.1 | - | 6 | 422 | |||
AK167162.1 | - | 7 | 659 | |||
AK168212.1 | - | 6 | 445 | |||
AK168706.1 | - | 6 | 405 | |||
AK168766.1 | - | 6 | 439 | |||
U31934.1 | - | 5 | 356 | |||
U76416.1 | - | 6 | 452 | |||
U82627.1 | - | 6 | 461 | |||
U94402.1 | - | 6 | 512 | |||
X97575.1 | - | 6 | 393 | |||
X99739.1 | - | 6 | 440 |

More Information
Other Species Targeted

Gene Ontology Categories:
