siRNA Details
Gene Symbol: Trim43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034906.2 | NP_001030078.1 | 3 | 629 | ||
GenBank mRNA | AK145647.1 | - | 3 | 630 | ||
AK166678.1 | - | 3 | 628 | |||
BC141236.1 | - | 3 | 508 | 9055848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
