siRNA Details
Gene Symbol: Ugt2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024148.1 | NP_001019319.1 | 2 | 673 | ||
GenBank mRNA | BC048920.1 | - | 1 | 674 | ||
BC058786.1 | - | 2 | 673 | 6310060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
