siRNA Details
Gene Symbol: Cypt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191032.1 | NP_001177961.1 | 1, 2 | 161 | ||
GenBank mRNA | BC099555.1 | - | 1, 2 | 161 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cypt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191032.1 | NP_001177961.1 | 1, 2 | 161 | ||
GenBank mRNA | BC099555.1 | - | 1, 2 | 161 |