siRNA Details
Gene Symbol: Tmem253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033805.3 | NP_001028977.1 | 7 | 1186 | ||
GenBank mRNA | AK144313.1 | - | 7 | 1187 |

More Information
Other Species Targeted

Gene Ontology Categories:
