siRNA Details
Gene Symbol: Defb42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034910.3 | NP_001030082.1 | 3 | 328 | ||
GenBank mRNA | AK079042.1 | - | 3 | 329 | ||
BC146022.1 | - | 2 | 184 | 40131325 | ||
BC146024.1 | - | 2 | 184 | 40131327 | ||
DQ012046.1 | - | 3 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
