siRNA Details
Gene Symbol: Vmn2r116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104580.1 | NP_001098050.1 | 3, 4 | 1304 | ||
GenBank mRNA | AB540947.1 | - | 3, 4 | 1304 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
