siRNA Details
Gene Symbol: Vmn2r117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104581.1 | NP_001098051.1 | 3 | 1189 | ||
GenBank mRNA | AB540948.1 | - | 3 | 1189 |
Gene Symbol: Vmn2r114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102584.1 | NP_001096054.1 | 3 | 1189 | ||
GenBank mRNA | AB540945.1 | - | 3 | 1189 |
Gene Symbol: Vmn2r115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104579.1 | NP_001098049.1 | 3 | 1189 | ||
GenBank mRNA | AB540946.1 | - | 3 | 1189 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
