siRNA Details
Gene Symbol: Igh-VJ558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF154882.2 | - | 141 | |||
AF296425.1 | - | 1 | 165 | |||
U38851.1 | - | 1 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
