siRNA Details
Gene Symbol: Bcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009740.2 | NP_033870.1 | 2 | 297 | ||
GenBank mRNA | AF057701.1 | - | 2 | 181 | ||
AF100339.1 | - | 2 | 189 | |||
AF127387.1 | - | 2 | 100 | |||
AF134396.1 | - | 2 | 181 | |||
AJ006289.1 | - | 2 | 178 | |||
AK076082.1 | - | 2 | 209 | |||
AK140179.1 | - | 2 | 169 | |||
AK146236.1 | - | 2 | 203 | |||
AK150883.1 | - | 2 | 198 | |||
AK152563.1 | - | 2 | 198 | |||
AK152847.1 | - | 2 | 194 | |||
AK156890.1 | - | 2 | 251 | |||
AK168084.1 | - | 2 | 203 | |||
AK169126.1 | - | 2 | 204 | |||
AK172158.1 | - | 2 | 208 | |||
BC003703.1 | - | 2 | 217 | |||
BC024379.1 | - | 2 | 175 | 4976147 | ||
BC172007.1 | - | 2 | 181 | 9054293 | ||
BC172009.1 | - | 2 | 181 | 9054295 | ||
BU707261.1 | - | 2 | 297 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_031328, AB016069, BC061772.
Gene Ontology Categories:
