siRNA Details
Gene Symbol: Zfp963 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200023.1 | NP_001186952.1 | 4 | 726 | ||
GenBank mRNA | AK162670.1 | - | 4 | 727 | ||
BC037669.1 | - | 2 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
