siRNA Details
Gene Symbol: Gm13889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145034.1 | NP_001138506.1 | 1 | 1173 | ||
GenBank mRNA | AK143447.1 | - | 1 | 1124 | ||
BB560177.2 | - | 1 | 562 |

More Information
Other Species Targeted

