siRNA Details
Gene Symbol: C87414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164284.1 | NP_001157756.1 | 5 | 754 | ||
NM_001164285.1 | NP_001157757.1 | 3 | 607 | |||
GenBank mRNA | AK136012.1 | - | 3 | 564 | ||
AK139835.1 | - | 5 | 804 | |||
AK163343.1 | - | 687 | ||||
BC080704.1 | - | 4 | 712 | 30476802 | ||
BC083101.1 | - | 3 | 563 | 30477931 | ||
BC085232.1 | - | 5 | 687 | 30476183 |
Gene Symbol: Gm3259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270456.1 | NP_001257385.1 | 4 | 688 | ||
GenBank mRNA | DV658954.1 | - | 2 | 214 |
Gene Symbol: Gm6367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054302.1 | - | 562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
