siRNA Details
Gene Symbol: Gm6164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167153.1 | NP_001160625.1 | 1 | 376 |
Gene Symbol: Gm16442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167148.1 | NP_001160620.1 | 1 | 376 |
Gene Symbol: Gm16451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167149.1 | NP_001160621.1 | 1 | 376 |
Gene Symbol: Gm4133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167158.1 | NP_001160630.1 | 1 | 376 |
Gene Symbol: Gm4187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167159.1 | NP_001160631.1 | 1 | 376 |
Gene Symbol: Gm5725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166711.1 | NP_001160183.1 | 1 | 376 |
Gene Symbol: Gm5726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167146.1 | NP_001160618.1 | 1 | 376 |
Gene Symbol: Gm8653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167152.1 | NP_001160624.1 | 1 | 376 |
Gene Symbol: Gm8677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167147.1 | NP_001160619.1 | 1 | 376 |
Gene Symbol: Gm8720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166756.1 | NP_001160228.1 | 1 | 376 |
Gene Symbol: Vmn1r101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166836.1 | NP_001160308.1 | 1 | 376 |
Gene Symbol: Vmn1r117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166743.1 | NP_001160215.1 | 1 | 376 |
Gene Symbol: Vmn1r119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166708.1 | NP_001160180.1 | 1 | 376 |
Gene Symbol: Vmn1r129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166725.1 | NP_001160197.1 | 1 | 376 |
Gene Symbol: Vmn1r149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166840.1 | NP_001160312.1 | 1 | 376 |
Gene Symbol: Vmn1r158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166841.1 | NP_001160313.1 | 1 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
