siRNA Details
Gene Symbol: Gm6251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101561.2 | NP_001095031.2 | 1 | 536 | ||
GenBank mRNA | CN721608.1 | - | 1 | 536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
