siRNA Details
Gene Symbol: Gm4305 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166639.1 | NP_001160111.1 | 535 |
Gene Symbol: Gm4301 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166637.1 | NP_001160109.1 | 1 | 538 | ||
GenBank mRNA | BC147487.1 | - | 1 | 538 | 9056243 |
Gene Symbol: Gm4302 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166634.1 | NP_001160106.1 | 1 | 537 | ||
GenBank mRNA | BC158099.1 | - | 1 | 537 | 9007384 |
Gene Symbol: Gm4303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166638.1 | NP_001160110.1 | 1 | 535 | ||
GenBank mRNA | BC158090.1 | - | 1 | 535 | 9007379 |
Gene Symbol: Gm4307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166641.1 | NP_001160113.1 | 535 |
Gene Symbol: Gm4312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166636.1 | NP_001160108.1 | 1 | 538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
