siRNA Details
Gene Symbol: Gm9785 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147682.1 | - | 1 | 625 | 9056655 | |
BC147695.1 | - | 1 | 625 | 9056668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
