siRNA Details
Gene Symbol: Cox6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053071.2 | NP_444301.1 | 3 | 288 | ||
GenBank mRNA | AK012602.1 | - | 3 | 205 | ||
AK013459.1 | - | 3 | 204 | |||
BC024666.1 | - | 3 | 219 | 4195595 | ||
BC094413.1 | - | 3 | 191 | 1397127 | ||
BY100624.1 | - | 3 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
