siRNA Details
Gene Symbol: Rhox3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114157.1 | NP_001107629.1 | 6 | 1009 |
Gene Symbol: Rhox3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194063.3 | NP_918952.2 | 6 | 1009 |
Gene Symbol: Rhox3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102457.4 | NP_001095927.3 | 4 | 789 | ||
GenBank mRNA | BC087875.1 | - | 3 | 614 | ||
BC092527.1 | - | 3 | 433 |
Gene Symbol: Rhox3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040089.3 | NP_001035178.3 | 4 | 789 | ||
GenBank mRNA | AY147207.1 | - | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
