siRNA Details
Gene Symbol: Gm6297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK139286.1 | - | 1 | 977 | ||
AK139295.1 | - | 1 | 509 | |||
AK172127.1 | - | 1 | 524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
