siRNA Details
Gene Symbol: Smok3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039889.3 | NP_001034978.1 | 2 | 713 | ||
GenBank mRNA | AK133180.1 | - | 2 | 714 |
Gene Symbol: Smok2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013741.1 | NP_038769.1 | 2 | 348 | ||
GenBank mRNA | AJ245453.1 | - | 2 | 348 |
Gene Symbol: Smok2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167913.2 | NP_001161385.1 | 2 | 682 |
Gene Symbol: Smok3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126045.1 | NP_001119517.1 | 2 | 539 | ||
NM_001168602.1 | NP_001162073.1 | 2 | 692 | |||
GenBank mRNA | AJ245454.1 | - | 2 | 692 | ||
BC150960.1 | - | 539 | 9087871 |
Gene Symbol: Smok4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029728.1 | - | 2 | 699 | ||
BC125398.1 | - | 1 | 257 | |||
BC132092.1 | - | 1 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
