siRNA Details
Gene Symbol: Fcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028632.2 | NP_082908.2 | 3 | 121 | ||
GenBank mRNA | AK003561.1 | - | 2 | 80 | ||
BC033045.1 | - | 2 | 99 | 1068150 | ||
BC094619.1 | - | 3 | 108 | |||
CJ085020.1 | - | 3 | 136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
