siRNA Details
Gene Symbol: Cks1brt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037922.3 | NP_001033011.1 | 2 | 517 | ||
GenBank mRNA | AK144293.1 | - | 2 | 518 | ||
BC147100.1 | - | 1 | 414 | 9056144 | ||
BC147101.1 | - | 1 | 414 | 9056154 |
Gene Symbol: Cks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016904.1 | NP_058600.1 | 2 | 250 | ||
GenBank mRNA | AB025409.1 | - | 2 | 250 | ||
AK004101.1 | - | 2 | 236 | |||
AK010414.2 | - | 2 | 237 | |||
AK011314.1 | - | 2 | 239 | |||
AK011782.1 | - | 2 | 216 | |||
BC076579.1 | - | 2 | 242 | 6846517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
