siRNA Details
Gene Symbol: 4930555G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175393.4 | NP_780602.2 | 5 | 1401 | ||
GenBank mRNA | AK076920.1 | - | 5 | 1402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
