siRNA Details
Gene Symbol: Taf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025444.2 | NP_079720.1 | 4 | 315 | ||
GenBank mRNA | AK007227.1 | - | 4 | 321 | ||
AK007339.1 | - | 4 | 316 | |||
AK076595.1 | - | 315 | ||||
AK160171.1 | - | 4 | 315 | |||
BC021447.1 | - | 4 | 327 | 5050736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
