siRNA Details
Gene Symbol: Rsl24d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198609.2 | NP_941011.1 | 2, 3 | 234 | ||
GenBank mRNA | AK153492.1 | - | 2, 3 | 246 | ||
AK160194.1 | - | 2, 3 | 246 | |||
AK164856.1 | - | 2, 3 | 231 | |||
BC003885.1 | - | 2, 3 | 234 | 3590401 | ||
BC089481.1 | - | 2, 3 | 231 | 30254073 |

More Information
Other Species Targeted

Gene Ontology Categories:
