siRNA Details
Gene Symbol: Gm6460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037919.3 | NP_001033008.1 | 3 | 394 | ||
GenBank mRNA | BC110565.1 | - | 3 | 394 | 40050213 |
Gene Symbol: 4933402N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177510.1 | NP_001170981.1 | 3 | 394 | ||
GenBank mRNA | AK077088.1 | - | 459 | |||
BC133658.1 | - | 3 | 394 | 40050211 |
Gene Symbol: Gm5861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030042.1 | - | 375 |
Gene Symbol: Gm6455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC048648.1 | - | 3 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
