siRNA Details
Gene Symbol: Vmn2r81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175936.1 | NP_787950.1 | 1 | 86 | ||
GenBank mRNA | AJ543404.1 | - | 1 | 86 |
Gene Symbol: Vmn2r82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101572.1 | NP_001095042.1 | 1 | 61 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
