siRNA Details
Gene Symbol: Cks1brt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037922.3 | NP_001033011.1 | 2 | 454 | ||
GenBank mRNA | AK144293.1 | - | 2 | 455 | ||
BC147100.1 | - | 1 | 351 | 9056144 | ||
BC147101.1 | - | 1 | 351 | 9056154 |

More Information
Other Species Targeted

Gene Ontology Categories:
