siRNA Details
Gene Symbol: BC147527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037925.2 | NP_001033014.1 | 5 | 1181 | ||
GenBank mRNA | AK132621.1 | - | 5 | 1182 | ||
BC147527.1 | - | 4 | 882 | 9056519 | ||
BC147545.1 | - | 4 | 882 | 9056536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
