siRNA Details
Gene Symbol: Fam43b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081672.2 | NP_001075141.1 | 1 | 641 | ||
GenBank mRNA | AK134570.1 | - | 1 | 641 |

More Information
Other Species Targeted

Gene Ontology Categories:
