siRNA Details
Gene Symbol: RPL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000993.4 | NP_000984.1 | 3 | 271 | ||
NM_001098577.2 | NP_001092047.1 | 3 | 271 | |||
NM_001099693.1 | NP_001093163.1 | 3 | 271 | |||
GenBank mRNA | AK297167.1 | - | 3 | 208 | ||
AK297247.1 | - | 3 | 208 | |||
AK297483.1 | - | 3 | 207 | |||
AK311775.1 | - | 3 | 208 | |||
BC017343.2 | - | 2 | 186 | 4895396 | ||
BC070210.1 | - | 3 | 207 | 4714258 | ||
BC070373.1 | - | 1 | 207 | 4666695 | ||
BE564858.1 | - | 2 | 188 | |||
BG031488.1 | - | 2 | 188 | |||
BG223187.1 | - | 1 | 225 | |||
CD108396.1 | - | 1 | 179 | |||
CF124259.1 | - | 3 | 265 | |||
CN269912.1 | - | 2 | 151 | |||
DA869102.1 | - | 3 | 271 | |||
X15940.1 | - | 2 | 191 | |||
X69181.1 | - | 2 | 172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
