siRNA Details
Gene Symbol: Ino80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC061495.1 | - | 4236 |
Gene Symbol: Rpl35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130484.1 | NP_001123956.1 | 4 | 348 | ||
NM_001130485.1 | NP_001123957.1 | 4 | 332 | |||
NM_021338.3 | NP_067313.2 | 4 | 378 | |||
GenBank mRNA | AK003063.1 | - | 4 | 302 | ||
AK010692.1 | - | 4 | 299 | |||
AK028214.1 | - | 4 | 302 | |||
AK046386.1 | - | 4 | 304 | |||
AK160963.1 | - | 3 | 1440 | |||
AV486753.1 | - | 4 | 384 | |||
BC027223.1 | - | 4 | 347 | 4036717 | ||
BC090255.1 | - | 3 | 285 | 2655001 | ||
BC100332.1 | - | 269 | 30846392 | |||
BC100599.1 | - | 4 | 302 | 6762930 | ||
BU530423.1 | - | 4 | 339 | |||
CJ043213.1 | - | 4 | 349 | |||
Y16430.2 | - | 3 | 287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
