siRNA Details
Gene Symbol: Gm11981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076664.1 | - | 3 | 293 | ||
BB614940.1 | - | 3 | 293 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm11981 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076664.1 | - | 3 | 293 | ||
BB614940.1 | - | 3 | 293 |