siRNA Details
Gene Symbol: Gm14322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243903.1 | NP_001230832.1 | 5 | 2290 | ||
GenBank mRNA | BC092145.1 | - | 6 | 1384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
