siRNA Details
Gene Symbol: Zfp800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081678.1 | NP_001075147.1 | 5 | 1447 | ||
GenBank mRNA | AK036691.1 | - | 6 | 1714 | ||
BC066073.1 | - | 5 | 1859 | |||
BC119236.1 | - | 5 | 1447 | 8733985 | ||
BC119262.1 | - | 5 | 1447 | 8734011 | ||
BC144761.1 | - | 5 | 1447 | 9053309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
