siRNA Details
Gene Symbol: Xlr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045539.2 | NP_001039004.2 | 8, 9 | 761 | ||
GenBank mRNA | BC051681.1 | - | 8, 9 | 761 | 30021721 |
Gene Symbol: Xlr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111293.1 | NP_001104763.1 | 8, 9 | 716 |
