siRNA Details
Gene Symbol: Rps27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033865.1 | NP_001029037.1 | 5 | 758 | ||
NM_024277.2 | NP_077239.1 | 6 | 663 | |||
GenBank mRNA | AK018706.1 | - | 6 | 485 | ||
AK159742.1 | - | 1 | 122 | |||
BC002108.1 | - | 5 | 473 | 3487291 | ||
BC081446.1 | - | 5 | 464 | 6408051 | ||
BC096392.1 | - | 5 | 477 | 1380923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
