siRNA Details
Gene Symbol: Gm6760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177377.1 | NP_001170848.1 | 2 | 272 | ||
GenBank mRNA | BC048649.1 | - | 2 | 167 | ||
BU939216.1 | - | 2 | 273 | |||
CN841342.1 | - | 2 | 183 |
