siRNA Details
Gene Symbol: Gm6792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177416.1 | NP_001170887.1 | 1, 2 | 189 | ||
GenBank mRNA | AK079003.1 | - | 14 | |||
AY226991.1 | - | 1, 2 | 188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
