siRNA Details
Gene Symbol: Slc39a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172653.2 | NP_766241.2 | 9 | 2343 | ||
GenBank mRNA | AK030685.1 | - | 479 | |||
AK049099.1 | - | 7 | 1089 | |||
AK122483.1 | - | 8 | 1777 | |||
BC052880.1 | - | 9 | 2457 | 30045566 | ||
BC059214.1 | - | 7 | 1227 | |||
BC062918.1 | - | 9 | 2343 | 6833098 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
