siRNA Details
Gene Symbol: Gm14725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081476.1 | NP_001074945.1 | 1 | 693 |
Gene Symbol: 4930567H17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033807.2 | NP_001028979.1 | 1 | 705 | ||
GenBank mRNA | AK133075.1 | - | 1 | 706 |
