siRNA Details
Gene Symbol: Igkv14-126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB089683.1 | - | 1 | 239 | ||
AB089684.1 | - | 1 | 239 | |||
AB248085.1 | - | 1 | 239 | |||
AF045511.1 | - | 2 | 299 | |||
AF294226.1 | - | 1 | 209 | |||
AY498735.1 | - | 1 | 239 | |||
D00079.1 | - | 1 | 239 | |||
GU563192.1 | - | 1 | 215 | |||
JX420448.1 | - | 1 | 239 | |||
JX420452.1 | - | 1 | 239 | |||
JX420735.1 | - | 1 | 239 | |||
JX420737.1 | - | 1 | 239 | |||
JX420738.1 | - | 1 | 239 | |||
JX420739.1 | - | 1 | 239 | |||
JX420741.1 | - | 1 | 239 | |||
JX420754.1 | - | 1 | 239 | |||
JX420755.1 | - | 1 | 239 | |||
JX420756.1 | - | 1 | 239 | |||
JX420757.1 | - | 1 | 239 | |||
JX420758.1 | - | 1 | 239 | |||
JX420775.1 | - | 1 | 239 | |||
JX492347.1 | - | 1 | 224 | |||
KC906592.1 | - | 1 | 233 | |||
M14840.1 | - | 1 | 239 | |||
M14842.1 | - | 1 | 239 | |||
M14843.1 | - | 1 | 239 | |||
M14844.1 | - | 1 | 239 | |||
M14845.1 | - | 1 | 239 | |||
M31955.1 | - | 1 | 239 | |||
M34593.1 | - | 1 | 239 | |||
M36256.1 | - | 1 | 239 | |||
M59918.1 | - | 2 | 305 | |||
M59919.1 | - | 2 | 305 | |||
U12575.1 | - | 1 | 215 | |||
U54503.1 | - | 1 | 219 | |||
U54504.1 | - | 1 | 219 | |||
U54506.1 | - | 1 | 219 | |||
U54508.1 | - | 1 | 219 | |||
U54512.1 | - | 1 | 219 | |||
U54514.1 | - | 1 | 163 | |||
U54515.1 | - | 1 | 215 | |||
U54516.1 | - | 1 | 211 | |||
U54517.1 | - | 1 | 182 | |||
U54518.1 | - | 1 | 219 | |||
U54519.1 | - | 1 | 159 | |||
U54520.1 | - | 1 | 203 | |||
U54521.1 | - | 1 | 219 | |||
U54522.1 | - | 1 | 215 | |||
U54523.1 | - | 1 | 181 | |||
U54524.1 | - | 1 | 201 | |||
U54525.1 | - | 1 | 180 | |||
U54527.1 | - | 1 | 219 | |||
U54529.1 | - | 1 | 200 | |||
U54531.1 | - | 1 | 202 | |||
U54532.1 | - | 1 | 200 | |||
U54533.1 | - | 1 | 219 | |||
U54537.1 | - | 1 | 219 | |||
U54538.1 | - | 1 | 211 | |||
U54539.1 | - | 1 | 181 | |||
U54540.1 | - | 1 | 185 | |||
U54541.1 | - | 1 | 214 | |||
U54542.1 | - | 1 | 219 | |||
U54543.1 | - | 1 | 214 | |||
U54546.1 | - | 1 | 219 | |||
U54547.1 | - | 1 | 219 | |||
U54548.1 | - | 1 | 219 | |||
U54549.1 | - | 1 | 219 | |||
X12411.1 | - | 1 | 191 | |||
X53353.1 | - | 1 | 239 | |||
X53355.1 | - | 1 | 239 | |||
X53356.1 | - | 1 | 239 | |||
X75104.1 | - | 1 | 239 |
