siRNA Details
Gene Symbol: Vmn2r114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102584.1 | NP_001096054.1 | 4 | 1385 | ||
GenBank mRNA | AB540945.1 | - | 4 | 1385 |
Gene Symbol: Vmn2r111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104573.1 | NP_001098043.1 | 4 | 1388 | ||
GenBank mRNA | AB194094.1 | - | 2 | 418 | ||
AB540944.1 | - | 4 | 1388 |
Gene Symbol: Vmn2r112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104575.1 | NP_001098045.1 | 4 | 1388 | ||
GenBank mRNA | AB194093.1 | - | 2 | 418 | ||
AB540943.1 | - | 4 | 1388 |
Gene Symbol: Vmn2r115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104579.1 | NP_001098049.1 | 4 | 1388 | ||
GenBank mRNA | AB540946.1 | - | 4 | 1388 |
Gene Symbol: Vmn2r116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104580.1 | NP_001098050.1 | 4 | 1385 | ||
GenBank mRNA | AB540947.1 | - | 4 | 1385 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
