siRNA Details
Gene Symbol: Gm38425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251890.1 | NP_001238819.1 | 3 | 587 | ||
GenBank mRNA | DV646607.1 | - | 1 | 40 | ||
DV654665.1 | - | 3 | 587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
